Skip to Content
Merck
All Photos(1)

Documents

1.10988

Millipore

Pseudomonas agar P (base)

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

form

medium granules (dehydrated (DCM))

Quality Level

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Pseudomonas spp.

General description

for microbiology

Analysis Note

Appearance (clearness): clear
Appearance (colour): yellowish-brown
pH-value (25 °C): 7.0 - 7.4
Growth (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): good to very good
Growth (Pseudomonas aeruginosa ATCC 9027 (WDCM 00026)): good to very good
Growth (Pseudomonas aeruginosa ATCC 25668 (WDCM 00114)): good to very good
Growth (Pseudomonas fluorescens ATCC 13525 (WDCM 00115)): good to very good
Growth (Aeromonas hydrophila ATCC 7966 (WDCM 00063)): good to very good
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): good to very good
Growth (Enterobacter cloacae ATCC 13047 (WDCM 00083)): good to very good
Blue-green pigment (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): (+)
Blue-green pigment (Pseudomonas aeruginosa ATCC 9027 (WDCM 00026)): +
Blue-green pigment (Pseudomonas aeruginosa ATCC 25668 (WDCM 00114)): +
Blue-green pigment (Pseudomonas fluorescens ATCC 13525 (WDCM 00115)): -
Blue-green pigment (Aeromonas hydrophila ATCC 7966 (WDCM 00063)): -
Blue-green pigment (Escherichia coli ATCC 25922 (WDCM 00013)): -
Blue-green pigment (Enterobacter cloacae ATCC 13047 (WDCM 00083)): -
Incubation: 24 - 48 hrs.; 35 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service