Skip to Content
Merck
All Photos(1)

Documents

CCW4547

Millipore

Chromogenic Pseudomonas Agar Plates

new

Solid culture medium for selective isolation and detection of Pseudomonas aeruginosa, suitable for microbiology

Synonym(s):

CPA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606

Quality Level

form

solid

shelf life

2 mo. at 2‑14 °C (limited shelf life, expiry date on the label)

feature

ready-to-use

composition

agar, 10.00 g/L
buffers, 2.15 g/L
peptones, 14.00 g/L
selective chromogenic mix, 0.24 g/L

packaging

box of 30 plates

storage condition

(Tightly closed)

parameter

9 mL filling volume

plate diam.

55 mm

color

yellowish

pH

7.1±0.2 (25 °C)

application(s)

food and beverages
microbiology

suitability

Pseudomonas spp.

General description

Chromogenic Pseudomonas Agar Plates are ready-to-use media plates with solid culture medium for selective isolation and detection of Pseudomonas spp. through membrane filtration. The Pseudomonas colonies appear blue-green in color due to the presence of the selective chromogenic mixture in the medium.Filter the prepared sample through a 0.45 µm pore membrane and apply it onto the surface of the Chromogenic agar.

Application

Chromogenic Pseudomonas Agar Plates are ready-to-use media plates suitable for selective isolation of Pseudomonas spp. from the samples.

Components

1 box containing: 6 plastic bags with 5 plates of 55 mm/bag.

Other Notes

Each laboratory must evaluate the results according to their specifications. Presumptive isolation of Pseudomonas spp. must be confirmed by further microbiological and biochemical tests.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service