1.10648
Carbonate Hardness Test Strips, colorimetric
water hardness 4-24 °d, MQuant®
Synonym(s):
Carbonate Hardness chemical test strips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
Carbonate Hardness Test, colorimetric, water hardness 4-24 °d, MQuant®
description
semi-quantitative
Quality Level
product line
MQuant®
specific analyte(s)
carbonate, hydrogen carbonate
water hardness measuring range
4-24 °d
detection method
colorimetric
storage temp.
15-25°C
General description
Method: Colorimetric, with test strips, 4 - 8 - 12 - 16 - 24°d, MQuant®
The MQuant® Carbonate hardness test strips are suitable for a rapid determination of carbonate hardness. Carbonate hardness (“temporary hardness”, alkalinity) is that proportion of hardness ions for which there is an equivalent amount of anions of carbonic acid (hydrogen carbonate and carbonate). As an alternative to testing for carbonate hardness with a test strip, one can also use a titration method. By using MQuant® titration tests (e.g. 111103) lower concentrations can be measured with finer graduation.
The MQuant® Carbonate hardness test strips are suitable for a rapid determination of carbonate hardness. Carbonate hardness (“temporary hardness”, alkalinity) is that proportion of hardness ions for which there is an equivalent amount of anions of carbonic acid (hydrogen carbonate and carbonate). As an alternative to testing for carbonate hardness with a test strip, one can also use a titration method. By using MQuant® titration tests (e.g. 111103) lower concentrations can be measured with finer graduation.
Linkage
Replaces: 10648-1; 10648
Legal Information
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service