Skip to Content
Merck
All Photos(1)

Documents

1.14653

Supelco

Carbonate Hardness Test Kit, titrimetric

titrimetric, for use with MQUANT®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Carbonate Hardness Test in freshwater and seawater, titrimetric, for use with MQUANT®

specific analyte(s)

carbonate, hydrogen carbonate

Quality Level

compatibility

for use with MQUANT®

detection method

titrimetric

storage temp.

15-25°C

General description

Carbonate hardness is a measure of the buffering capacity of water which can also be expressed as acid-binding capacity or total alkalinity. This MQuant® Carbonate Hardness test (formally MColortest®) is suitable for the titrimetric determination using a convenient dropping bottle.

Method
Hydrogen carbonate and carbonate ions are titrated with hydrochloric acid against a mixed indicator. At the titration end-point (at pH 4.3) the color changes to red. The carbonate hardness (acid capacity) is determined from the consumption of titration solution.
Method: titrimetric with dropping bottle MQuant®
The Carbonate Hardness Test determines carbonate hardness as a measure of the buffering capacity of water which can also be expressed as acid-binding capacity or total alkalinity.

Analysis Note

Application test: conforms

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service