Skip to Content
Merck
All Photos(1)

Key Documents

1.06023

Millipore

TAE buffer

10 x pH 8.3 tris-acetate-EDTA buffer

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
1.060,00 kr.

1.060,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 L
1.060,00 kr.

About This Item

UNSPSC Code:
41105319
NACRES:
NA.25

1.060,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

form

liquid

pH

8.2-8.4 ( in H2O)

density

1.016 g/cm3 at 20 °C

storage temp.

15-25°C

General description

10 x pH 8.3 tris-acetate-EDTA buffer

Analysis Note

Appearanceclear and colourlesspH-value8.2 - 8.4DNases (Exo- and endonucleases)not detectableRNasesnot detectableProteasesnot detectableComposition:
Tris(hydroxymethyl)-aminomethane 0.40 mol/l
EDTA 0.01 mol/l
Acetic acid 0.20 mol/l

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service