Skip to Content
Merck
All Photos(2)

Documents

855175P

Avanti

06:0 Lyso PC

1-hexanoyl-2-hydroxy-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-hexanoyl-sn-glycero-3-phosphocholine; PC(6:0/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H30NO7P
CAS Number:
Molecular Weight:
355.36
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 25 mg (855175P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855175P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCC)=O

InChI

1S/C14H30NO7P/c1-5-6-7-8-14(17)20-11-13(16)12-22-23(18,19)21-10-9-15(2,3)4/h13,16H,5-12H2,1-4H3/t13-/m1/s1

InChI key

WDNDPXJAUNUOFK-CYBMUJFWSA-N

General description

06:0 Lyso PC is a natural lysophosphatidylcholine.

Packaging

5 mL Clear Glass Sealed Ampule (855175P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R E Stafford et al.
Biochemistry, 28(12), 5113-5120 (1989-06-13)
The critical micelle concentration (cmc) of several lysophospholipids and of a lysophospholipid analogue was determined from surface tension measurements using the maximum bubble pressure method and/or 31P NMR. The use of the maximum bubble pressure method has now been extended

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service