Skip to Content
Merck
All Photos(1)

Documents

878126P

Avanti

C18:1 Lyso PAF

Avanti Research - A Croda Brand 878126P, powder

Synonym(s):

1-O-(9Z)octadecenyl-2-hydroxy-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H54NO6P
CAS Number:
Molecular Weight:
507.68
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (878126P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 878126P

lipid type

bioactive lipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COCCCCCCCC/C=C\CCCCCCCC

Application

C18:1 Lyso PAF, also known as 1-O-(9Z)octadecenyl-2-hydroxy-sn-glycero-3-phosphocholine is suitable for evaluating substrate specificity of P.putida KT2440 lyPlsase.

Packaging

5 mL Clear Glass Sealed Ampule (878126P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advances in Clinical Chemistry (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service