Key Documents
DWK07-2000
Wheaton® Microliter μlplate Medium Polypropylene
Select a Size
1 EACH
77,60 kr.
77,60 kr.
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
77,60 kr.
About This Item
UNSPSC Code:
41106200
NACRES:
NB.15
material:
polypropylene plate
round bottom
round wells
77,60 kr.
Please contact Customer Service for Availability
material
polypropylene plate
round bottom
round wells
packaging
pkg of 1 ea
manufacturer/tradename
WHEATON® 07-2000
capacity
500 μL
Application
Chromatography
Features and Benefits
A comprehensive line of 96-well microplates for chromatography, the Plate Sampling System uses closures familiar to those who work with standard chromatography vials. Polypropylene μLplates have a variety of closures that will stabilize the sample during the analytical sample run and, if necessary, reseal after the sample has been injected. You may also choose the security of a stopper coupled with the strength of a mechanical closure system. The mechanical closure system permits even GC samples can be converted to 96-well format, by sealing the limited volume insert on the inside surface of the insert and compressing the PTFE/silicone rubber using a mechanical seal. A variety of insert options let you choose the specifications appropriate for your application. The system combines microplates, glass inserts, μLmats, and caps with chromatography-based septa to offer you secure vessels to maintain even the most volatile samples.
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)