Skip to Content
Merck
All Photos(1)

Key Documents

244309

Sigma-Aldrich

Ethyl β-carboline-3-carboxylate

97%

Synonym(s):

β-CCE, Ethyl 9H-pyrido[3,4-b]indole-3-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N2O2
CAS Number:
Molecular Weight:
240.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

228-230 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)c1cc2c(cn1)[nH]c3ccccc23

InChI

1S/C14H12N2O2/c1-2-18-14(17)12-7-10-9-5-3-4-6-11(9)16-13(10)8-15-12/h3-8,16H,2H2,1H3

InChI key

KOVRZNUMIKACTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Yasui et al.
Neuropharmacology, 32(2), 127-131 (1993-02-01)
The effects of benzodiazepine inverse agonists on the long-term potentiation of synaptic transmission in hippocampal slices of the guinea pig were examined using an extracellular recording technique. Benzodiazepine inverse agonists, beta-carboline-3-carboxylate (beta-CCE), 2-phenylpyrazolo [4,3-c]quinolin-3(5H)-one (CGS-8216) and 2-[5-methylthien-3-yl]-2,5-dihydro-3H-pyrazolo [4,3-c]quinolin-3-one (S-135), augmented
L R Gerak et al.
Pharmacology, biochemistry, and behavior, 61(4), 375-380 (1998-11-05)
This study examined changes in ventilation produced by negative gamma-aminobutyric acid(A) (GABA(A)) modulators in rhesus monkeys. The effects of Ro 15-4513, beta-CCE and beta-CCM were examined in four rhesus monkeys breathing air or 5% CO2 in air. When monkeys breathed
Beta-carboline-3-carboxylic acid ethyl ester: a lead for new psychotropic drugs.
R Schmiechen et al.
Psychopharmacology series, 11, 7-15 (1993-01-01)
B K Koe et al.
European journal of pharmacology, 90(1), 97-102 (1983-05-20)
Ethyl beta-carboline-3-carboxylate (beta CCE), a benzodiazepine antagonist, was found to increase basal levels of cyclic GMP in rat cerebellum. beta CCE also augmented the elevation of cyclic GMP concentrations induced by isoniazid, in contrast to diazepam which blocked this effect
M Katsura et al.
European journal of pharmacology, 298(1), 71-77 (1996-02-29)
Effect of exposure of primary cultured cerebral cortical neurons to ethyl beta-carboline-3-carboxylate (beta-CCE) on the function of benzodiazepine receptors was studied. Exposure of neurons to beta-CCE (0.1-10 microM) decreased the binding of [3H]flunitrazepam to extensively washed membrane fractions in a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service