Skip to Content
Merck
All Photos(1)

Key Documents

240257

Sigma-Aldrich

Thiourea

ACS reagent

Synonym(s):

Sulfourea, Thiocarbamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CSNH2
CAS Number:
Molecular Weight:
76.12
Beilstein:
605327
EC Number:
MDL number:
UNSPSC Code:
12352100

Pricing and availability is not currently available.

grade

ACS reagent

ign. residue

≤0.1%

loss

≤0.5% loss on drying

mp

170-176 °C (lit.)

SMILES string

NC(N)=S

InChI

1S/CH4N2S/c2-1(3)4/h(H4,2,3,4)

InChI key

UMGDCJDMYOKAJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chaotropic agent; strong denaturant. Increases solubility and recovery of proteins
Used in the formation of thiols from alkyl halides.

replaced by

Product No.
Description
Pricing

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service