11020
DL-Arginine
≥95% (NT)
Synonym(s):
(±)-2-Amino-5-guanidinopentanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HN=C(NH2)NH(CH2)3CH(NH2)COOH
CAS Number:
Molecular Weight:
174.20
Beilstein:
1725411
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥95% (NT)
reaction suitability
reaction type: solution phase peptide synthesis
application(s)
peptide synthesis
SMILES string
NC(CCCNC(N)=N)C(O)=O
InChI
1S/C6H14N4O2/c7-4(5(11)12)2-1-3-10-6(8)9/h4H,1-3,7H2,(H,11,12)(H4,8,9,10)
InChI key
ODKSFYDXXFIFQN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jurgen R Piet et al.
The Journal of infectious diseases, 209(11), 1781-1791 (2013-12-18)
Streptococcus pneumoniae (pneumococcus) is a major human pathogen causing pneumonia, sepsis and bacterial meningitis. Using a clinical phenotype based approach with bacterial whole-genome sequencing we identified pneumococcal arginine biosynthesis genes to be associated with outcome in patients with pneumococcal meningitis.
Lindsay D Plank et al.
Hepatology (Baltimore, Md.), 61(2), 639-647 (2014-09-13)
Preliminary work suggested that perioperative immunonutrition (IMN) enriched in n-3 fatty acids, arginine, and nucleotides may improve preoperative nutritional status, enhance postoperative recovery, and reduce postoperative infectious complications in patients undergoing liver transplantation (LT). The current study examined these outcomes
Gaël Piton et al.
Shock (Augusta, Ga.), 44(5), 438-444 (2015-07-22)
Cardiac arrest is considered to be a cause of small bowel ischemia, but the consequences of cardiac arrest on the human small bowel have been rarely studied. Plasma citrulline concentration is a marker of functional enterocyte mass, and plasma intestinal
Phyllis S Y Chong et al.
Cancer research, 74(11), 3043-3053 (2014-04-02)
PRL-3, an oncogenic dual-specificity phosphatase, is overexpressed in 50% of acute myelogenous leukemia (AML) and associated with poor survival. We found that stable expression of PRL-3 confers cytokine independence and growth advantage of AML cells. However, how PRL-3 mediates these
Isabelle Dunand-Sauthier et al.
Journal of immunology (Baltimore, Md. : 1950), 193(4), 1690-1700 (2014-07-11)
Arginine, a semiessential amino acid implicated in diverse cellular processes, is a substrate for two arginases-Arg1 and Arg2-having different expression patterns and functions. Although appropriately regulated Arg1 expression is critical for immune responses, this has not been documented for Arg2.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service