Skip to Content
Merck
All Photos(1)

Key Documents

8.14584

Sigma-Aldrich

4-Propylcyclohexanone

for synthesis

Synonym(s):

4-Propylcyclohexanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O
CAS Number:
Molecular Weight:
140.22
MDL number:
UNSPSC Code:
12352115
EC Index Number:
406-810-4
NACRES:
NA.22

Pricing and availability is not currently available.

vapor pressure

0.23 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

320 °C

potency

>2000 mg/kg LD50, oral (Rat)

pH

6.0 (20 °C, 2 g/L in H2O)

bp

219 °C/1013 hPa

mp

<-70 °C

transition temp

flash point 90 °C

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.906 - 0.908
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service