Merck
  • Muscle Satellite Cell Cross-Talk with a Vascular Niche Maintains Quiescence via VEGF and Notch Signaling.
Muscle Satellite Cell Cross-Talk with a Vascular Niche Maintains Quiescence via VEGF and Notch Signaling.
Cell stem cell (2018-10-06)
Mayank Verma, Yoko Asakura, Bhavani Sai Rohit Murakonda, Thomas Pengo, Claire Latroche, Benedicte Chazaud, Linda K McLoon, Atsushi Asakura
ABSTRACT
Skeletal muscle is a complex tissue containing tissue resident muscle stem cells (satellite cells) (MuSCs) important for postnatal muscle growth and regeneration. Quantitative analysis of the biological function of MuSCs and the molecular pathways responsible for a potential juxtavascular niche for MuSCs is currently lacking. We utilized fluorescent reporter mice and muscle tissue clearing to investigate the proximity of MuSCs to capillaries in 3 dimensions. We show that MuSCs express abundant VEGFA, which recruits endothelial cells (ECs) in vitro, whereas blocking VEGFA using both a vascular endothelial growth factor (VEGF) inhibitor and MuSC-specific VEGFA gene deletion reduces the proximity of MuSCs to capillaries. Importantly, this proximity to the blood vessels was associated with MuSC self-renewal in which the EC-derived Notch ligand Dll4 induces quiescence in MuSCs. We hypothesize that MuSCs recruit capillary ECs via VEGFA, and in return, ECs maintain MuSC quiescence though Dll4.
Sigma-Aldrich
MISSION® esiRNA, targeting human DLL1
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Acetamide, ~99% (GC)
Sigma-Aldrich
Diatrizoic acid, Iodine-containing contrast agent
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biology
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
Boric acid, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
N-Methyl-D-glucamine, 99.0-100.5% (titration)
Sigma-Aldrich
Histodenz, nonionic density gradient medium
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Barium chloride, 99.9% trace metals basis
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)