Skip to Content
Merck
All Photos(1)

Key Documents

96607

Supelco

2-Phenylindole

analytical standard

Synonym(s):

NSC 15776, Stabilizer I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11N
CAS Number:
Molecular Weight:
193.24
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥ 98.0% (GC)

technique(s)

HPLC: suitable

bp

250 °C/10 mmHg (lit.)

mp

188-190 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

c1ccc(cc1)-c2cc3ccccc3[nH]2

InChI

1S/C14H11N/c1-2-6-11(7-3-1)14-10-12-8-4-5-9-13(12)15-14/h1-10,15H

InChI key

KLLLJCACIRKBDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition
  • Tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
  • Organic light emitting diodes (OLEDs)
  • Anti inflammatory agents
  • Potential cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors
  • Agents for cancer and malaria therapy
  • Antifungal and antibacterial agents
  • Fluorescent probes

Reactant in:
  • Difluorohydroxylation reactions†
  • Mannich-type reactions†

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service