Skip to Content
Merck
All Photos(1)

Key Documents

52913

Supelco

Hexatetracontane

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€59.60

€59.60


In StockDetails


Request a Bulk Order

Select a Size

Change View
250 MG
€59.60

About This Item

Linear Formula:
CH3(CH2)44CH3
CAS Number:
Molecular Weight:
647.24
Beilstein:
1805968
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€59.60


In StockDetails


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

86-91 °C

application(s)

petroleum

format

neat

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C46H94/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39-41-43-45-46-44-42-40-38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-46H2,1-2H3

InChI key

PVQAQPUODMPGGG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service