Documents
509590
Syringes Fitted with Luer Lock Valve (509353)
volume 1000 μL, 1MDR-VLL-GT, needle size (not included)
About This Item
UNSPSC Code:
41122001
material
glass syringe
needle size
(not included)
feature
Pre-fitted with Luer Lock Valve
packaging
pkg of 1 ea
manufacturer/tradename
SGE 008160
volume
1000 μL
Related Categories
General description
  • SGE Syringes (volume 1 mL, needle size (not included)) fitted with Luer Lock Valve, Model: 1MDR-VLL-GT.
  • Made using borosilicate glass, PTFE plunger tips, and KEL-F Luer cones.
  • Autoclavable syringes but the plunger needs to be removed.
  • Feature an interference fit between the plunger tip and the barrel, that improves the gas sealing properties of the syringe.
  • Maximum inertness for liquid or gas samples.
Choose from one of the most recent versions:
Certificates of Analysis (COA)