Documents
22285
Push Button Valve
for use with Hamilton TLL syringes
About This Item
UNSPSC Code:
41122001
material
PTFE Mininert valve
packaging
pkg of 1 ea
manufacturer/tradename
654051
VICI Precision Sampling
compatibility
for use with Hamilton TLL syringes
Related Categories
General description
This Mininert valve will attach to your syringe and provide sample storing capability. It features push-button operation and an all-PTFE body with a stainless steel miniature valve stem.
Application
Push button valve has been used in headspace analysis of volatile compounds in equilibrium partitioning in closed systems (EPICS) bottles using GC coupled with laboratory computing integrator. It has also been used in gas sampling procedure for analysing gas compositions in hypertrophic lake.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Measurement of Henry's law constants for C1 and C2 chlorinated hydrocarbons.
Gossett, James M.
Environmental Science & Technology, 21.2, 202-208 (1987)
Fluxes of methane and carbon dioxide from a small productive lake to the atmosphere.
Casper, Peter, et al.
Biogeochemistry, 49.1, 1-19 (2000)