Skip to Content
Merck
All Photos(8)

Key Documents

ZRB2282

Sigma-Aldrich

Anti-DNAJB9 Antibody, clone 1D16-K ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

ER-resident protein ERdj4, Endoplasmic reticulum DNA J domain-containing protein 4, Microvascular endothelial differentiation gene 1 protein, UNQ743/PRO1471

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D16-K, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 25.52 kDa
observed mol wt ~25 kDa

purified by

using Protein A

species reactivity

human, mouse, rat

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... DNAJB9(4189)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1D16-K is a ZooMAb® rabbit recombinant monoclonal antibody that detects DNAJB9. It targets an epitope within 23 amino acids from the C-terminal half of the protein.

Immunogen

KLH-conjugated linear peptide corresponding to 23 amino acids from the C-terminal half of human DNAJB9.

Application

Quality Control Testing

Evaluated by Western Blotting in mouse kidney tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected DNAJB9 in mouse kidney tissue lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected DNAJB9 in rat kidney tissue lysates.

Affinity Binding Assay: The binding affinity of this antibody to DNAJB9 peptide was at least one hundred thousand-fold (100,000X) higher than that obtained with the non-specific control peptide.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected DNAJB9 in human kidney and mouse kidney tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

DnaJ homolog subfamily B member 9 (UniProt: Q9UBS3; also known as Endoplasmic reticulum DNA J domain-containing protein 4, ER-resident protein ERdj4, Microvascular endothelial differentiation gene 1 protein) is encoded by the DNAJB9 (also known as MDG1, UNQ743/PRO1471) gene (Gene ID:4189) in human. DNAJB9 belongs to the DnaJ/Hsp40 protein family, which is involved in chaperone-mediated protein folding and cellular stress responses. DNAJB9 consists of various functional domains, including a J domain, which is responsible for interacting with Hsp70 chaperones, and a cysteine-rich domain implicated in protein-protein interactions. The expression profile of DNAJB9 varies across different tissues and cell types. It is expressed in various organs, including the brain, heart, liver, and kidney. DNAJB9 expression levels can be regulated under different physiological and pathological conditions, such as during cellular stress or in certain disease states. Increased expression of DNAJB9 has been observed in certain brain regions affected by diseases. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service