Z743994
Worthington LD Series Liquid Nitrogen Dewars
LD50, beaker style, capacity 50 L
Synonym(s):
Liquid Nitrogen Storage Dewar
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
beaker style
packaging
pack of 1 ea
manufacturer/tradename
Worthington 50LDB
technique(s)
cryopreservation: suitable
H × D
32.4 in. × 18.8 in.
capacity
50 L
neck size
2.5 in. , diameter
weight
38.7 lb
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The LD Series Liquid Nitrogen Dewars are designed to store and dispense small amounts of liquid nitrogen.
- Provides high thermal efficiency
- Ribbed, high-strength aluminum body, magniformed neck tube design, and durable paint make these dewars rugged
- Easy to operate design features a lightweight snap-on cap and necktube provides a tight closure and easy access
- Provides maximum holding times with superior vacuum performance and insulation
- Accessories sold separately
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service