Skip to Content
Merck
All Photos(1)

Key Documents

U5133

Sigma-Aldrich

Anti-Ubiquitin C-terminal Hydrolase L1 (MA-18) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-UCH-L1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 27 kDa

species reactivity

human, rat, mouse

technique(s)

microarray: suitable
western blot: 1:1,000-1:2,000 using cytosolic fraction (S1) of mouse brain or whole cell extract of human lung carcinoma A549 cell line
western blot: 1:10,000-1:20,000 using cytosolic fraction (S1) of rat brain

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UCHL1(7345)
mouse ... Uchl1(22223)
rat ... Uchl1(29545)

Immunogen

synthetic peptide corresponding to amino acids 1-18 located at the N-terminus of rat UCH-L1, conjugated to KLH. This sequence is identical in human, mouse, bovine, porcine, and guinea pig UCH-L1. No homology is found with other known UCH-L isoforms.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service