Skip to Content
Merck
All Photos(1)

Documents

K2894

Sigma-Aldrich

PKAcβ, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

cAPKb

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

423-573 nmol/min·mg

mol wt

~65 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRKACB(5567)

Biochem/physiol Actions

PRKACB (protein kinase cAMP-activated catalytic subunit β) is a catalytic subunit allele of PKA (protein kinase A). Mutation in it is associated with neural tube defects. PRKACB can phosphorylate p65 in presence of cyclic adenosine monophosphate (cAMP).

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seiko Susuki-Miyata et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(14), E1800-E1809 (2015-04-02)
Phosphodiesterase 4B (PDE4B) plays a key role in regulating inflammation. Roflumilast, a phosphodiesterase (PDE)4-selective inhibitor, has recently been approved for treating severe chronic obstructive pulmonary disease (COPD) patients with exacerbation. However, there is also clinical evidence suggesting the development of
J Simard et al.
Human genetics, 88(6), 653-657 (1992-03-01)
A cDNA for the human catalytic subunit (C beta) of cAMP-dependent protein kinase (PKA) has been cloned from a testis cDNA library. In the present study, we have determined the chromosomal localization of this gene using a cDNA for C
Jian Wu et al.
International journal of clinical and experimental pathology, 6(12), 2968-2974 (2013-12-03)
This study aimed to investigate the single nucleotide polymorphisms (SNPs) of PKA and neural tube defects (NTDs) in Chinese population. A total of 183 NTDs cases and 200 healthy controls were used in this study. 7 selected single nucleotide polymorphism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service