Skip to Content
Merck
All Photos(3)

Key Documents

HPA003265

Sigma-Aldrich

Anti-FGF1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Acidic fibroblast growth factor, Anti-Beta-endothelial cell growth factor, Anti-HBGF-1, Anti-Heparin-binding growth factor 1 precursor, Anti-aFGF

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

GEITTFTALTEKFNLPPGNYKKPKLLYCSNGGHFLRILPDGTVDGTRDRSDQHTDTK

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FGF1(2246)

General description

The gene FGF1 (fibroblast growth factor 1) is mapped to human chromosome 5q31. The protein encoded by this gene lacks signal peptides and is released from damaged cells. It may also be released by an exocytotic mechanism that is not related to the ER-Golgi pathway. The FGF gene family in humans is divided into seven subfamilies.

Immunogen

Heparin-binding growth factor 1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

FGF1 (fibroblast growth factor 1) gene encodes a protein belonging to the fibroblast growth factor family of proteins that are essential for mammalian development. It is involved in the mesodermal induction, neurulation, and the formation of the circulatory and skeletal systems during embryogenesis. FGF1 and FGF2 function in several biological processes including angiogenesis, regeneration of tissue, inflammation and the formation of some tumors.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85210.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Igor Prudovsky et al.
Journal of cell science, 116(Pt 24), 4871-4881 (2003-11-20)
Non-classical protein release independent of the ER-Golgi pathway has been reported for an increasing number of proteins lacking an N-terminal signal sequence. The export of FGF1 and IL-1alpha, two pro-angiogenic polypeptides, provides two such examples. In both cases, export is
Nobuyuki Itoh et al.
Trends in genetics : TIG, 20(11), 563-569 (2004-10-12)
Fibroblast growth factors (Fgfs) and Fgf receptors (Fgfrs) comprise a signaling system that is conserved throughout metazoan evolution. Twenty-two Fgfs and four Fgfrs have been identified in humans and mice. During evolution, the Fgf family appears to have expanded in

Articles

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service