Skip to Content
Merck
All Photos(4)

Key Documents

MAB4212F

Sigma-Aldrich

Anti-HU-B27 Antibody, FITC conjugated

Chemicon®, from mouse

Synonym(s):

Com-B27

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€459.00

€459.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
1 ML
€459.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€459.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

clone

monoclonal

species reactivity

human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG2a

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

HU-B27 (HLA- Com-B27) is involved in the presentation of foreign antigens to the immune system. HLA B belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta 2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from the endoplasmic reticulum lumen. They are expressed in nearly all cells.

The previously assigned protein identifier P03989 has been merged into P01889. Full details can be found on the UniProt database.

Specificity

This reagent binds strongly to the HLA-B27 antigen. It also binds weakly to the HLAB7 antigen. This formulation demonstrates less cross reactivity to HLA-B7 than Chemicon products, MAB1285 and MAB1285F.

Application

Detect HU-B27d using this Anti-HU-B27 Antibody, FITC conjugated validated for use in FC, IC, IF & WB.
Flow cytometry and Immunofluorescence: use 10 mL direct from the vial per 100 mL of whole blood, or 1 x 106 peripheral blood mononuclear cells (PBMC) in 100 mL buffer.

Flow cytometric analysis of lymphocyte staining for HLA-B27 gave the following results:
Subjects/Fluorescence Intensity
Heterozygous HLA-B27+/Strong
Homozygous HLA-B7+/Weak
Heterozygous HLA-B7+/Weak - negative
Non B27, Non B7/Negative

This reagent is useful in the investigation of HLA-B27 in diseases such as ankylosing spondylitis.

SUGGESTED USAGE:
Flow cytometry and immunofluorescence - use 10 µl direct from the vial per 100 µl of whole blood, or 1 x 10E6 peripheral blood mononuclear cells (PBMC) in 100 µl buffer.

Use Control IgG2a-FITC (MABC002F) as an isotype control.

Please note that this reagent should be used in a single-colour single-step assay only.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by Western Blot on Jurkat lysates.

Western Blotting Analysis:
1:500 dilution of this antibody detected Com-B27 on 10 µg of Jurkat lysates.

Target description

19 kDa

Physical form

The reagent is supplied in 1.0 mL phosphate-buffered saline, pH 7.4, containing 0.2% bovine serum albumin and 0.1% sodium azide. Purified from mouse ascitic fluid. The characteristics of each lot are tested by gel electrophoresis and flow cytometry.

Storage and Stability

Stable for 6 months at 2-8ºC in the dark from date of receipt.

Analysis Note

Control
Control IgG2a-FITC (MABC002F) as an isotype control

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Description of a mouse monoclonal anti-HLA-B27 antibody HLA-ABC-m3.
Trapani, J A, et al.
Human immunology, 7, 205-216 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service