Skip to Content
Merck
All Photos(1)

Documents

GE28-9889-49

Sigma-Aldrich

XK 26/40 Column

Synonym(s):

XK Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
NACRES:
SB.52

technique(s)

LPLC: suitable

Related Categories

General description

XK empty columns provide reliable and reproducible separations. The tube material is of borosilicate glass, which allows for visual inspection of the media bed. The columns are made of biocompatible materials that exhibit excellent chemical resistance. A broad range of column dimensions makes XK columns suitable for many different purification needs. Column packing can be performed using either a packing reservoir or an extra column tube attached with a packing connector. The QuickLock mechanism of the adapter shaft facilitates rapid and easy movement of the adapter, simplifying adjustments of the bed height, disassembly, and cleaning. The design of the adapter plunger gives a uniform flow, which maintains the integrity of the packed bed during operations. An extensive range of spare parts is available, facilitating maintenance.

Features and Benefits

Adapter QuickLock mechanism that facilitates column handling and cleaning
Cooling jacket for sensitive purifications
Fittings for direct connection to chromatographiy systems
Plunger design to ensure a homogeneous buffer distribution and reproducible packing
User friendly and robust empty columns, designed for liquid chromatography at low to medium pressure.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This page covers how and why to use Sepharose Big Beads for purification from crude, viscous samples.

Here we discuss purification options when working with Sepharose® High Performance and we present some examples of media screening, capture and purification.

This page shows how to purify histidine-tagged proteins using TALON Superflow from Cytiva.

This page shows how to separate IgG antibodies by affinity chromatography using Protein G Sepharose 4 Fast Flow from Cytiva.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service