Skip to Content
Merck
All Photos(1)

Documents

GE28-4110-07

HiTrap® Hic Selection Kit

Cytiva 28-4110-07, pack of 7 × 1 mL

Synonym(s):

HIC selection kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

packaging

pack of 7 × 1 mL

manufacturer/tradename

Cytiva 28-4110-07

parameter

42 psi

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

Related Categories

General description

HiTrap® HIC Selection Kit consists of seven 1 mL columns prepacked with hydrophobic interaction chromatography media with different characteristics. Screen for the most appropriate HIC media and conditions to use for specific application and development work.

Application

For screening to find the best HIC medium for a specific application.

Features and Benefits

  • Select the most appropriate ligand and experimental conditions to take full advantage of hydrophobic interaction chromatography.
  • Flexible design for use with syringes, single laboratory pump, or chromatography system such as AKTA design.
  • High flow rates and high capacities

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

HiTrap is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page provides information about the principles and standard conditions for different purification techniques for the purification of multiprotein complexes with from Cytiva.

Column, media, and sample preparation for Hydrophobic Interaction Chromatography (HIC) detailed with Cytiva media.

This page describes principles and standard conditions for different purification techniques of histidine-tagged proteins using Cytiva products.

Protocols

Here we discuss purification options when working with Sepharose® High Performance and we present some examples of media screening, capture and purification.

This page discusses various aspects of sample preparation for chromatographic purification.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service