Skip to Content
Merck
All Photos(1)

Key Documents

EHU024581

Sigma-Aldrich

MISSION® esiRNA

targeting human PDCD6IP

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€194.00
50 μG
€343.00

€194.00


Estimated to ship on29 May 2025



Select a Size

Change View
20 μG
€194.00
50 μG
€343.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€194.00


Estimated to ship on29 May 2025


description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCAAAGCCACACTTGTGAAATCTACCCCGGTCAATGTACCCATCAGTCAGAAATTTACTGATCTGTTTGAGAAGATGGTTCCCGTGTCAGTACAGCAGTCTTTGGCTGCCTATAATCAGAGGAAAGCCGATTTGGTTAACAGATCAATTGCTCAGATGAGAGAAGCCACCACTTTGGCAAATGGGGTGCTAGCTTCCCTTAATCTTCCAGCAGCAATTGAAGATGTGTCTGGAGACACTGTACCTCAGTCTATATTGACTAAATCCAGATCTGTGATTGAACAGGGAGGCATCCAGACTGTTGATCAGTTGATTAAAGAACTGCCTGAATTACTGCAACGAAATAGAGAAATCCTAGATGAGTCATTAAGGTTGTTGGATGAAGAAGAAGCAACCGATAATGATTTAAGAGCAAAATTTAAGGAACGTTGGCAAAGGACACCATCCAATGAACTGTATAAGCCTTTAAGAGCAGAGGGAACCAACTTCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julianne V de Carvalho et al.
PloS one, 9(11), e113691-e113691 (2014-11-26)
Nef is an HIV-1 accessory protein that promotes viral replication and pathogenesis. A key function of Nef is to ensure sustained depletion of CD4 and MHC-I molecules in infected cells by inducing targeting of these proteins to multivesicular bodies (MVBs)
Li Zhong et al.
Signal transduction and targeted therapy, 6(1), 59-59 (2021-02-12)
It remains unknown for decades how some of the therapeutic fusion proteins positive in a small percentage of cancer cells account for patient outcome. Here, we report that osteosarcoma Rab22a-NeoF1 fusion protein, together with its binding partner PYK2, is sorted
Allaura S Cone et al.
BMC molecular and cell biology, 21(1), 58-58 (2020-08-01)
Endosomal trafficking and amyloidogenic cleavage of amyloid precursor protein (APP) is believed to play a role in the neurodegeneration observed in Alzheimer's disease (AD). Recent evidence has suggested that packaging and secretion of APP and its amyloidogenic cleaved products into
Brian A Davies et al.
Molecular biology of the cell, 31(22), 2463-2474 (2020-08-28)
Intercellular communication is critical for organismal homeostasis, and defects can contribute to human disease states. Polarized epithelial cells execute distinct signaling agendas via apical and basolateral surfaces to communicate with different cell types. Small extracellular vesicles (sEVs), including exosomes and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service