Skip to Content
Merck
All Photos(1)

Key Documents

AV34909

Sigma-Aldrich

Anti-CHRNA1 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Cholinergic receptor, nicotinic, α-1 (muscle)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€423.00

€423.00


Estimated to ship on14 April 2025



Select a Size

Change View
100 μL
€423.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€423.00


Estimated to ship on14 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

52 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHRNA1(1134)

General description

CHRNA1 encodes the α subunit of the muscle acetylcholine receptor and is involved in acetyl choline binding and channel gating functions. The expression of CHRNA1 in thymus tissues is controlled by IRF8 and AIRE.
Rabbit Anti-CHRNA1 recognizes mouse, bovine, canine, and human CHRNA1.

Immunogen

Synthetic peptide directed towards the N terminal region of human CHRNA1

Application

Rabbit Anti-CHRNA1 is suitable for western blot applications at a concentration of 1.8 μg/ml.

Biochem/physiol Actions

The muscle acetylcholine receptor consiststs of 5 subunits of 4 different types: 2 alpha isoforms and 1 each of beta, gamma, and delta subunits.2 CHRNA1 encodes an alpha subunit that plays a role in acetlycholine binding/channel gating.

Sequence

Synthetic peptide located within the following region: TRLVAKLFKDYSSVVRPVEDHRQVVEVTVGLQLIQLINVDEVNQIVTTNV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthieu Giraud et al.
Nature, 448(7156), 934-937 (2007-08-10)
Promiscuous expression of tissue-restricted auto-antigens in the thymus imposes T-cell tolerance and provides protection from autoimmune diseases. Promiscuous expression of a set of self-antigens occurs in medullary thymic epithelial cells and is partly controlled by the autoimmune regulator (AIRE), a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service