Skip to Content
Merck
All Photos(1)

Key Documents

A1005

Sigma-Aldrich

(+)-6-Aminopenicillanic acid

Synonym(s):

(2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-APA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2O3S
CAS Number:
Molecular Weight:
216.26
Beilstein:
15080
EC Number:
MDL number:
UNSPSC Code:
12352200

mp

198-200 °C (dec.) (lit.)

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

CC1(C)S[C@@H]2[C@H](N)C(=O)N2[C@H]1C(O)=O

InChI

1S/C8H12N2O3S/c1-8(2)4(7(12)13)10-5(11)3(9)6(10)14-8/h3-4,6H,9H2,1-2H3,(H,12,13)/t3-,4+,6-/m1/s1

InChI key

NGHVIOIJCVXTGV-ALEPSDHESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junqi Zhao et al.
Bioresource technology, 102(2), 529-535 (2010-10-23)
In this study, macro-mesoporous silica spheres were prepared with a micro-device and used as the support for the immobilization of penicillin G acylase (PGA). To measure the enzymatic activity, the silica spheres with immobilized PGA were placed into a packed-bed
Ye-Wang Zhang et al.
Applied microbiology and biotechnology, 88(1), 49-55 (2010-06-23)
A one-pot, two-step enzymatic synthesis of amoxicillin from penicillin G, using penicillin acylase, is presented. Immobilized penicillin acylase from Kluyvera citrophila was selected as the biocatalyst for its good pH stability and selectivity. Hydrolysis of penicillin G and synthesis of
Joanna Skiba et al.
European journal of medicinal chemistry, 57, 234-239 (2012-10-18)
We report on the synthesis of ferrocenyl-ampicillin and ferrocenyl-6-aminopenicillinic acid bioconjugates. Title compounds were characterized by (1)H NMR, IR, MS and elemental analysis. These novel ferrocenyl-antibiotic conjugates were also investigated by cyclic voltammetry (CV). Ferrocenyl-ampicillin complexes revealed reversible uncomplicated oxidation
A K Dolui et al.
Indian journal of experimental biology, 49(4), 289-292 (2011-05-28)
In the present study different bacterial samples were isolated from soil of different places of Dibrugarh and screened for biotransformation ability to produce 6-Aminopenicillanic acid. Among ten isolated bacterial samples, three gram positive bacterial samples designated as AKDD-2, AKDD-4 and
Louis Anthony Tony Cox et al.
Risk analysis : an official publication of the Society for Risk Analysis, 29(6), 796-805 (2009-06-06)
Penicillin and ampicillin drugs are approved for use in food animals in the United States to treat, control, and prevent diseases, and penicillin is approved for use to improve growth rates in pigs and poultry. This article considers the possibility

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service