Merck
  • Immobilization of penicillin G acylase on macro-mesoporous silica spheres.
Immobilization of penicillin G acylase on macro-mesoporous silica spheres.
Bioresource technology (2010-10-23)
Junqi Zhao, Yujun Wang, Guangsheng Luo, Shenlin Zhu
ABSTRACT
In this study, macro-mesoporous silica spheres were prepared with a micro-device and used as the support for the immobilization of penicillin G acylase (PGA). To measure the enzymatic activity, the silica spheres with immobilized PGA were placed into a packed-bed reactor, in which the hydrolysis of penicillin G was carried out. The influences of the residence time, the initial concentration of the substrate, the accumulation of the target product 6-aminopenicillanic acid, and the enzyme loading amount on the performance of the immobilized PGA were investigated. The introduction of macropores increased the enzyme loading amount and decreased the internal mass transfer resistance, and the results showed that the enzyme loading amount reached 895 mg/g (dry support), and the apparent enzymatic activity achieved up to 1033 U/g (dry support). In addition, the immobilized PGA was found to have great stability.
Sigma-Aldrich
(+)-6-Aminopenicillanic acid, 96%