Skip to Content
Merck
All Photos(1)

Documents

IRMM618

Rubidium-87Rb solution

IRMM®, certified reference material, 0.5 M in nitric acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

IRMM®

isotopic purity

 87Rb

manufacturer/tradename

JRC

concentration

0.5 M in nitric acid

application(s)

general analytical

format

single component solution

storage temp.

10-30°C

General description

Isotope abundance ratio

Analysis Note

For more information please see:
IRMM618

Legal Information

IRMM is a registered trademark of European Commission

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service