Skip to Content
Merck
All Photos(1)

Documents

NIST3129A

Lithium (Li) standard solution

NIST® SRM® 3129a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

format

single component solution

General description

One unit of SRM 3129a comprises five 10 mL sealed borosilicate glass ampoules. Each ampoule contains a solution prepared gravimetrically with a known mass fraction of lithium. The solution also includes nitric acid at a concentration of approximately 0.16 mol/L. For more information, please refer to the SDS and COA.

SRM 3129A_cert

SRM 3129A_SDS

Application

Lithium (Li) standardsolution SRM is intended for use as a primary calibrationstandard for the quantitative determination of lithium.

Features and Benefits

  • NIST certified value of Lithiumis metrologically traceable to the SI unit for mass fraction and expressed asmilligrams per gram and determined using gravimetric preparation usinghigh-purity lithium carbonate and inductively coupled plasma optical emissionspectrometry (ICP-OES).
  • Expiration details, safety, storage, and useare provided on the NIST certificate.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Lithium (Li)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Flame over circleCorrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Ox. Liq. 3 - Skin Irrit. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service