C100
Flow Cup Viscosity Standard
UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 281.4 mPa.s (20 °C)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
viscosity
281.4 mPa.s(20 °C)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
mixture
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Flow Cup Viscosity Standards can be found on the Paragon Scientific webpage
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sajjad Esmaeili et al.
International journal of biological macromolecules, 91, 368-380 (2016-05-24)
Ferritin is a giant protein composed of 24 subunits which is able to sequester up to 4500 atoms of iron. We proposed two kinds of heme binding sites in mammalian ferritins and provided direct evidence for peroxidase activity of heme-ferritin
Evan Kazura et al.
Biomedical microdevices, 19(3), 50-50 (2017-06-01)
Point of care (POC) diagnostics represents one of the fastest growing health care technology segments. Developments in microfabrication have led to the development of highly-sensitive nanocalorimeters ideal for directly measuring heat generated in POC biosensors. Here we present a novel
Verena Biswenger et al.
PloS one, 13(9), e0203040-e0203040 (2018-09-14)
Chemotactic cell migration is a central mechanism during cancer cell invasion and hence metastasis. In order to mimic in vivo conditions, we used a three-dimensional hydrogel matrix made of collagen I and a stable gradient-generating chemotaxis assay system, which is
Fabrício Motteran et al.
The Science of the total environment, 580, 1120-1128 (2017-01-04)
This study presents a new method developed for the simultaneous determination of anionic surfactant (linear alkylbenzene sulfonate - LAS, 4 homologs) and nonionic surfactant (linear alcohol ethoxylate - LAE) in commercial laundry wastewater. The surfactants were identified and quantified using
Brian J McHugh et al.
PLoS pathogens, 15(4), e1007694-e1007694 (2019-04-13)
Pulmonary infections are a major global cause of morbidity, exacerbated by an increasing threat from antibiotic-resistant pathogens. In this context, therapeutic interventions aimed at protectively modulating host responses, to enhance defence against infection, take on ever greater significance. Pseudomonas aeruginosa
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service