Skip to Content
Merck
All Photos(1)

Key Documents

VISRT500600

Silicone Rotational Viscosity Standard

503.9 mPa.s (25.00°C), RT500

Sign Into View Organizational & Contract Pricing

Select a Size

600 ML
€196.00

€196.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
600 ML
€196.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€196.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

ASTM D 2162

503.9 mPa.s(25.00 °C)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service