Skip to Content
Merck
All Photos(1)

Key Documents

92564

Supelco

p-Acetamidophenyl β-D-glucuronide sodium salt

analytical standard

Synonym(s):

Acetaminophen glucuronide, Paracetamol glucuronide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C14H16NO8Na
CAS Number:
Molecular Weight:
349.27
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥97% (TLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[Na+].CC(=O)Nc1ccc(O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C([O-])=O)cc1

InChI

1S/C14H17NO8.Na/c1-6(16)15-7-2-4-8(5-3-7)22-14-11(19)9(17)10(18)12(23-14)13(20)21;/h2-5,9-12,14,17-19H,1H3,(H,15,16)(H,20,21);/q;+1/p-1/t9-,10-,11+,12-,14+;/m0./s1

InChI key

OINXIJJEOMGKPB-CYRSAHDMSA-M

Looking for similar products? Visit Product Comparison Guide

Application

p-Acetamidophenyl β-D-glucuronide sodium salt has been used as a reference standard for the evaluation of paracetamol drug metabolism in human plasma and urine samples by high performance liquid chromatography coupled to tandem mass spectrometry (HPLC–MS/MS) method.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous quantitative determination of paracetamol and its glucuronide conjugate in human plasma and urine by liquid chromatography coupled to electrospray tandem mass spectrometry: application to a clinical pharmacokinetic study.
Tan Q Y, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 893, 162-167 (2015)
Andrew Rowland et al.
Drug metabolism and disposition: the biological fate of chemicals, 43(1), 147-153 (2014-11-09)
This study characterized the kinetics, variability, and factors that affect UDP-glucuronic acid (UDP-GlcUA) uptake by human liver microsomes (HLM). Biphasic kinetics were observed for UDP-GlcUA uptake by HLM. Uptake affinities (assessed as Kd) of the high- and low-affinity components differed
Paula Paíga et al.
Journal of pharmaceutical and biomedical analysis, 106, 61-70 (2014-07-09)
An analytical methodology for the simultaneous determination of seven pharmaceuticals and two metabolites belonging to the non-steroidal anti-inflammatory drugs (NSAIDs) and analgesics therapeutic groups was developed based on off-line solid-phase extraction and ultra-high performance liquid chromatography coupled to tandem mass

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service