Skip to Content
Merck
p-Acetamidophenyl β-D-glucuronide sodium salt >98% (TLC)

p-Acetamidophenyl β-D-glucuronide sodium salt

p-Acetamidophenyl β-D-glucuronide sodium salt >98% (TLC)
Synonym(s):
Acetaminophen glucuronide, Paracetamol glucuronide
Linear Formula:
C14H16NO8Na
CAS Number:
Molecular Weight:
349.27

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing