Skip to Content
Merck
All Photos(1)

Key Documents

18-1400

Sigma-Aldrich

Lithium carbonate

SAJ first grade, ≥98.0%

Synonym(s):

Carbolithium, Carbonic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2CO3
CAS Number:
Molecular Weight:
73.89
Beilstein:
3999191
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥98.0%
grade:
SAJ first grade
form:
crystalline

grade

SAJ first grade

Assay

≥98.0%

form

crystalline

availability

available only in Japan

pH

9.0-11.0 (1 g/L)

mp

618 °C (lit.)

SMILES string

[Li+].[Li+].[O-]C([O-])=O

InChI

1S/CH2O3.2Li/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

XGZVUEUWXADBQD-UHFFFAOYSA-L

Gene Information

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Contestabile et al.
The Journal of clinical investigation, 123(1), 348-361 (2012-12-04)
Down syndrome (DS) patients exhibit abnormalities of hippocampal-dependent explicit memory, a feature that is replicated in relevant mouse models of the disease. Adult hippocampal neurogenesis, which is impaired in DS and other neuropsychiatric diseases, plays a key role in hippocampal
Tom Bschor et al.
Journal of clinical psychopharmacology, 33(1), 38-44 (2013-01-02)
Studies of the 1970s and 1980s showed lithium monotherapy to be an effective treatment of acute unipolar major depressive disorder (MDD) and hence as a potential alternative to monoaminergic antidepressants.The objective was to conduct the first comparison of a lithium
Andrew A Nierenberg et al.
The American journal of psychiatry, 170(1), 102-110 (2013-01-05)
Lithium salts, once the mainstay of therapy for bipolar disorder, have tolerability issues at a higher dosage that often limit adherence. The authors investigated the comparative effectiveness of more tolerable dosages of lithium as part of optimized personalized treatment (OPT).
M Simard et al.
Archives of internal medicine, 149(1), 36-46 (1989-01-01)
Lithium carbonate is a widely used and invaluable drug in the treatment and prevention of manic-depressive illness. However, this medication has a low therapeutic index and, therefore, many attendant side effects. Acute lithium carbonate intoxication affects predominantly the central nervous
R Yassa et al.
International pharmacopsychiatry, 15(5), 301-308 (1980-01-01)
In this paper, the efficacy of lithium therapy in the treatment of tardive dyskinesia is reviewed. It is noted that most of the published papers on this topic are anecdotal and single case reports with contradictory results. The well-designed studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service