Merck
  • Lithium carbonate intoxication. A case report and review of the literature.
Lithium carbonate intoxication. A case report and review of the literature.
Archives of internal medicine (1989-01-01)
M Simard, B Gumbiner, A Lee, H Lewis, D Norman
ABSTRACT
Lithium carbonate is a widely used and invaluable drug in the treatment and prevention of manic-depressive illness. However, this medication has a low therapeutic index and, therefore, many attendant side effects. Acute lithium carbonate intoxication affects predominantly the central nervous system and the renal system and is potentially lethal. We studied a case of acute lithium carbonate intoxication characterized by a prolonged comatose state, severe nephrogenic diabetes insipidus, and glucose intolerance. Of interest, the patient's symptoms and signs of toxic reaction developed after normalization of the plasma lithium ion levels. We reviewed the multisystemic toxic effects of lithium carbonate as well as its teratogenic potential and guidelines for its use during pregnancy. The management of acute lithium carbonate intoxication should include hydration with normal saline, intensive care monitoring, and dialysis if indicated.
Sigma-Aldrich
Lithium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Lithium carbonate, 99.997% trace metals basis
Sigma-Aldrich
Lithium carbonate, puriss. p.a., ACS reagent, reagent (for microscopy), ≥99.0% (T)
Sigma-Aldrich
Lithium carbonate, ACS reagent, ≥99.0%