Skip to Content
Merck
All Photos(1)

Key Documents

MAB5478

Sigma-Aldrich

Anti-Retinoid X Receptor α Antibody

ascites fluid, Chemicon®

Synonym(s):

RXRalpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
electrophoretic mobility shift assay: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RXRA(6256)

Specificity

Reacts with Retinoid X Receptor alpha (RXRalpha). No cross reactivity with RXRbeta or RXRgamma.

Immunogen

Fusion protein corresponding to the DE-region of mouse RXRa.

Application

Use Anti-Retinoid X Receptor α Antibody (Mouse Monoclonal Antibody) validated in ELISA, EMSA, IP, WB to detect Retinoid X Receptor α also known as RXR alpha.
Western blot (endogenous and recombinant protein): 1:500-1:5,000

Immunocytochemistry (transfected cells): 1:500-1:5,000

Immunoprecipitation (endogenous and recombinant protein): 1:500-1:5,000

Gel shift (endogenous and recombinant protein): 1:500-1:5,000

ELISA: 1:500-1:5,000

Optimal working dilutions must be determined by end user.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Retinoid-X-receptors (?/?) in melanocytes modulate innate immune responses and differentially regulate cell survival following UV irradiation.
Coleman, DJ; Garcia, G; Hyter, S; Jang, HS; Chagani, S; Liang, X; Larue, L; Ganguli-Indra et al.
PLoS Genetics null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service