Skip to Content
Merck
All Photos(1)

Documents

IEVH08100

Millipore

Immobilon® -E PVDF Membrane

10 sheets, 8 cm x 10 cm, 0.45 µm pore size, transfer membrane wets out in transfer buffer or water

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
eCl@ss:
32031602
NACRES:
NB.23

product name

Immobilon®-E PVDF Membrane, 10 sheets, 8 cm x 10 cm, The first PVDF transfer Membrane for western blotting that wets out in transfer buffer or water. For chemiluminescence or chromogenic detection.

material

PVDF membrane
plain filter
white filter

Quality Level

feature

easily wetted with water

manufacturer/tradename

Immobilon®

technique(s)

western blot: suitable

filter L × W

8 cm × 10 cm

pore size

0.45 μm pore size

compatibility

for use with Amido black
for use with Colloidal gold
for use with Coomassie brilliant blue
for use with India ink
for use with Ponceau-S red
for use with Sypro<TMSYMBOL></TMSYMBOL> ruby
for use with Transillumination

detection method

chemiluminescent
colorimetric
radioactive

shipped in

ambient

Related Categories

General description

Filter Code: IPVH
Primary Binding Mechanisms: Electrostatic, hydrophobic

Application

10 sheets, 8 cm x 10 cm, The first PVDF transfer Membrane for western blotting that wets out in transfer buffer or water. For chemiluminescence or chromogenic detection.

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service