Skip to Content
Merck
All Photos(1)

Key Documents

NWKITIE

Millipore

Immobilon® -E PVDF NOW Bundle

2 rolls, 8.5 cm x 10 m, 0.45 µm pore size, transfer membrane wets out in transfer buffer or water, 1 dispenser included

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NB.23
Pricing and availability is not currently available.

Product Name

Immobilon®-E NOW Bundle, The first PVDF transfer Membrane for western blotting that wets out in transfer buffer or water. For chemiluminescence or chromogenic detection.

material

PVDF membrane
plain filter
white filter

feature

easily wetted with water

packaging

dispenser box of 1
roll of 2

manufacturer/tradename

Immobilon®

technique(s)

western blot: suitable

roll W × L

8.5 cm × 10 m

pore size

0.45 μm pore size

detection method

chemiluminescent
colorimetric
radioactive

General description

Compatible stains:
  • Coomassie brilliant blue
  • Amido black
  • India ink
  • Ponceau-S red
  • Colloidal gold
  • Transillumination
  • Sypro ruby

Filter Code: IEVH
Immobilon-E Membrane is the only PVDF membrane that wets out in aqueous buffers, eliminating the methanol pre-wet step. It has a 45 μm pore size, is recommended for most Western blotting applications, especially proteins greater than 20 kDa, and is compatible with chemiluminescent and chromogenic detection.
Primary Binding Mechanisms: Electrostatic, hydrophobic
The Immobilon NOW system offers 1-cut convenience for standard mini-blots (7 x 8.5cm) and midi-blots (8.5 x 13.5cm).

Components

This bundle includes the Immobilon NOW Dispenser and two 8.5cm x 10m rolls of Immobilon-E transfer membrane.

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service