Skip to Content
Merck
All Photos(1)

Key Documents

8.20013

Sigma-Aldrich

Adamantane

for synthesis

Synonym(s):

Adamantane, Tricyclo[3.3.1.1(3,7)]decane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
MDL number:
UNSPSC Code:
12352001
EC Index Number:
206-001-4

Assay

≥99.0% (GC)

autoignition temp.

287 °C

mp

209-212 °C

storage temp.

2-30°C

InChI

1S/C10H16/c1-7-2-9-4-8(1)5-10(3-7)6-9/h7-10H,1-6H2

InChI key

ORILYTVJVMAKLC-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Identity (IR): passes test

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service