Skip to Content
Merck
All Photos(2)

Key Documents

8.18407

Sigma-Aldrich

(R)-(+)-Limonene

for synthesis

Synonym(s):

(R)-(+)-Limonene, (+)-p-Mentha-1,8-diene, (R)-(+)-4-Isopropenyl-1-methyl-1-cyclohexene

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€26.90
500 ML
€60.60

€26.90


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€26.90
500 ML
€60.60

About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
MDL number:
UNSPSC Code:
12352002
EC Index Number:
227-813-5
NACRES:
NA.22

€26.90


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

2.1 hPa ( 20 °C)

Quality Level

Assay

≥94.0% (GC)

form

liquid

autoignition temp.

255 °C

potency

>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.7-6.1 % (v/v)

bp

177-178 °C/1013 hPa

mp

-74 °C

transition temp

flash point 51 °C (DIN 51758)

density

0.84 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[C@H]1(CCC(=CC1)C)C(=C)C

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,10H,1,5-7H2,2-3H3/t10-/m0/s1

InChI key

XMGQYMWWDOXHJM-JTQLQIEISA-N

Application


  • Antioxidant activity of fermented orange waste: Explores the biotransformation of R-(+)-limonene in solid-state fermentation, leading to antioxidant compounds like limonene-1,2-diol (Bier et al., 2019).

  • Isomerization and dehydroaromatization: Investigates the catalytic activity of Ti-MCM-41 in the transformation of R-(+)-limonene, highlighting the catalyst′s efficiency and the influence of reaction conditions (Retajczyk and Wróblewska, 2019).

  • Activity against Anisakis larvae: Examines the potential use of R-(+)-limonene in seafood products to control Anisakis larvae, suggesting its effectiveness and application in food safety (Giarratana et al., 2015).

  • Biological effects of limonene types: Compares the effects of natural and synthetic sources of R-limonene on biological systems, focusing on its impact when used in various products (Nematollahi et al., 2021).

  • Limonene in Rhodosporidium toruloides engineering: Discusses the engineering of Rhodosporidium toruloides for enhanced limonene production, highlighting genetic modifications to increase yield (Liu et al., 2021).

Analysis Note

Assay (GC, area%): ≥ 94.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.841 - 0.846
Spec. rotation [α²0/D (undiluted): +115.0 - +122.0 °
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service