Skip to Content
Merck
All Photos(1)

Documents

05-1571

Sigma-Aldrich

Anti-IGF-1R Antibody, clone 24-57

clone 24-57, from mouse

Synonym(s):

CD221 antigen, IGF-I receptor, Insulin-like growth factor I receptor, insulin-like growth factor 1 receptor, soluble IGF1R variant 1, soluble IGF1R variant 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

24-57, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IGF1R(3480)

General description

There are two kinds of IGF receptors based on their relative affinities for IGF1 versus IGF2. The IGF1 receptor prefers IGF1 over IGF2 and weakly binds insulin. IGF1 receptor is a disulfide-linked heterotetrameric transmembrane protein consisting of two alpha (130 kDa) and two beta (95 kDa) subunits. Both the alpha and beta subunits are encoded within a single receptor precursor cDNA. The IGF1 receptor is therefore similar in structure to the insulin receptor. The proreceptor polypeptide is proteolytically cleaved and disulfide-linked to yield the mature heterotetrameric receptor. The IGF1 receptor is highly expressed in all cell types and tissues and is highly overexpressed in most malignant tissues where it functions as an anti-apoptotic agent by enhancing cell survival.

Specificity

This antibody recognizes IGF-IR.

Immunogen

Epitope: Unknown
KLH-conjugated linear peptide corresponding to human IGF-IR.

Application

Immunoprecipitation Analysis: A previous lot was used by an independent laboratory in IP. (Soos, M., et al. (1992). Journal of Biological Chemistry. 267(18):12955-12963.)
Research Category
Metabolism
Research Sub Category
Insulin/Energy Signaling
This Anti-IGF-1R Antibody, clone 24-57 is validated for use in IH, IP for the detection of IGF-1R.

Quality

Evaluated by Immunohistochemistry in human placental villi and tonsil tissue.

Immunohistochemistry Analysis: A 1:500 dilution from a previous lot detected IGF-1R in human placental villi and tonsil tissue.

Target description

155 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human placental villi and tonsil tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service