Z303097
Quickfit® air condenser
L 150 mm, joint: ST/NS 19/26
Synonym(s):
C2/12
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Joint
joint: ST/NS19/26
manufacturer/tradename
Quickfit C 2/12
availability
not available in USA and Canada, ROW (Available only in EU countries)
not available in USA and Canada
L
150 mm
surface area
100 cm2
Looking for similar products? Visit Product Comparison Guide
General description
These simple condensers are useful for condensation of materials with boiling points greater the 150 °C.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service