Skip to Content
Merck
All Photos(1)

Key Documents

MAB8241

Sigma-Aldrich

Anti-Hepatitis A Virus Antibody

Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€558.00

€558.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€558.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€558.00


Estimated to ship on02 June 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

Specificity

Hepatitis A virus. Specific for a neutralizing epitope. Does not react with related viruses.

Application

Anti-Hepatitis A Virus Antibody detects level of Hepatitis A Virus & has been published & validated for use in ELISA, IF, WB, IH.
IFA, IHC, Western Blot (unboiled samples in 1mM DTT and 2% SDS only): 1:10-1:50

ELISA: 1:20-1:200

Optimal working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.01 M phosphate buffered saline (PBS), pH 7.2 containing 0.1% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service