CLS2094250
Pyrex® colored scale dispensing burette, with straight bore PTFE stopcock plug
volume 250 mL, accuracy: 2.0 mL
Synonym(s):
burettes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
feature
subdivision 1 mL
manufacturer/tradename
Corning 2094-250
volume measuring range
250 mL, accuracy: 2.0 mL
volume
250 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Corning® 2094
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service