Skip to Content
Merck
All Photos(1)

Key Documents

88125

Sigma-Aldrich

Tetrapropylammonium (meta)periodate

moistened with water, ≥98.0% (calc. based on dry substance, RT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)4N(IO4)
CAS Number:
Molecular Weight:
377.26
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

≥98.0% (calc. based on dry substance, RT)

form

crystals

quality

moistened with water

contains

~10% water as stabilizer

reaction suitability

reagent type: oxidant

mp

181-183 °C (dec.) (lit.)

SMILES string

[O-]I(=O)(=O)=O.CCC[N+](CCC)(CCC)CCC

InChI

1S/C12H28N.HIO4/c1-5-9-13(10-6-2,11-7-3)12-8-4;2-1(3,4)5/h5-12H2,1-4H3;(H,2,3,4,5)/q+1;/p-1

InChI key

HDTMHCZUXQKRAJ-UHFFFAOYSA-M

Application

Reagent for:
  • Preparation of acylnitroso compoubnds used in Diels-Alder reactions

Other Notes

Oxidation reagent, soluble in organic solvents

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Supplementary Hazards

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G.E. Keck et al.
Tetrahedron, 37, 4007-4007 (1981)
G.E. Keck
Tetrahedron Letters, 4767-4767 (1978)
K. Inomata et al.
Bulletin of the Chemical Society of Japan, 53, 565-565 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service