Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Chem. Unst. Gas A - Eye Dam. 1 - Flam. Gas 1A - Muta. 1B - Press. Gas Liquefied gas - Repr. 1B - Skin Corr. 1B - STOT RE 1 - STOT SE 3
Target Organs
Central nervous system, Nervous system, Respiratory system
Storage Class Code
2A - Gases
WGK
WGK 3
Flash Point(F)
-4.0 °F - closed cup
Flash Point(C)
-20.0 °C - closed cup
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
Critical reviews in toxicology, 30(5), 595-608 (2000-10-31)
Long-term inhalation studies in rodents have presented unequivocal evidence of experimental carcinogenicity of ethylene oxide, based on the formation of malignant tumors at multiple sites. However, despite a considerable body of epidemiological data only limited evidence has been obtained of
A risk estimate of the heritable effects of ethylene oxide exposure, using the parallelogram approach, as suggested by Frits Sobels, is described. The approach is based on available data on the ethylene oxide-induced responses for the same genetic endpoint in
Data for relationships between in vivo doses inferred from levels of hemoglobin (Hb) or DNA adducts and administered (by inhalation or injection) doses of ethylene oxide (EO) in mice, rats and humans are reviewed. At low absorbed doses or dose
Allergy to formaldehyde and ethylene-oxide.
J Bousquet et al.
Clinical reviews in allergy, 9(3-4), 357-370 (1991-01-01)
Journal of dentistry, 42(8), 1027-1032 (2014-04-01)
To test the hypothesis that a material with higher water absorption than polyvinylidene fluoride (PVDF) could increase the yield of target molecules from exposed dentine. In a series of standard tests, different cellulose membranes were compared to a PVDF counterpart
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.