Skip to Content
Merck
All Photos(1)

Key Documents

537837

Sigma-Aldrich

2-Methyl-4-nitroanisole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)OCH3
CAS Number:
Molecular Weight:
167.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

61-64 °C (lit.)

functional group

nitro

SMILES string

COc1ccc(cc1C)[N+]([O-])=O

InChI

1S/C8H9NO3/c1-6-5-7(9(10)11)3-4-8(6)12-2/h3-5H,1-2H3

InChI key

QOZMIJZYJZQOBV-UHFFFAOYSA-N

General description

2-Methyl-4-nitroanisole is obtained as one of the products from the charge-transfer trinitromethylation of 2-methylanisole in the presence of dichloromethane.[1]

Application

2-Methyl-4-nitroanisole may be used in the synthesis of 2-methyl-4-nitrophenol[2] and 3-methyl-4-methoxyaniline.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

5-Methyl-6-methoxy-8-(2'-diethylaminoethylamino)-quinoline1.
Carmack M, et al.
Journal of the American Chemical Society, 68(8), 1551-1552 (1946)
Studies on photochemical reaction of air pollutants. X. Identification of nitrophenols in suspended particulates.
Nojima K, et al.
Chemical & Pharmaceutical Bulletin, 31(3), 1047-1051 (1983)
Annihilation of aromatic cation radicals by ion-pair and radical pair collapse. Unusual solvent and salt effects in the competition for aromatic substitution.
Sankararaman S, et al.
Journal of the American Chemical Society, 109(25), 7824- 7838 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service