Alkohole
Materialien
Standard
CONDITIONS
column
6 ft.x 2 mm I.D. glass
packing
5% Carbowax 20M on 60/80 Carbopack B
oven
85 °C
detector
FID
carrier gas
helium, 20 mL/min
injection
1 μL
sample
0.05-0.10% each analyte in water
Beschreibung
Rechtliche Hinweise
Carbopack is a trademark of Merck KGaA, Darmstadt, Germany
Carbowax is a registered trademark of Union Carbide